missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl (S)-(+)-3-nitrobenzenesulfonate, 98%

Product Code. p-7041687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441419 5 g 5g
15451419 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441419 Supplier Thermo Scientific Alfa Aesar Supplier No. H64289.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115314-14-2 | C9H9NO6S | 259.23 g/mol

Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycidyl (S)-(+)-3-nitrobenzenesulfonate is used in the synthesis of enantiopure bisoprolol by reacting with phenols using cesium fluoride as catalyst. It is also involved in the preparation of orally active HIV-1 protease inhibitor.

Solubility
Soluble in chloroform, methylene chloride and tetrahydrofuran. Insoluble in hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115314-14-2
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.23
MDL Number MFCD00064582
InChI Key AIHIHVZYAAMDPM-QMMMGPOBSA-N
Synonym s-+-glycidyl nosylate, s-glycidyl nosylate, s-glycidyl 3-nitrobenzenesulfonate, s-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, s-+-glycidyl 3-nitrobenzenesulfonate, glycidyl 3-nitrobenzenesulfonate, ccris 6394, 2s-+-glycidyl-3-nitrobenzenesulfonate, oxiranylmethyl s-3-nitrobenzenesulfonate, 2s-oxiran-2-yl methyl 3-nitrobenzenesulfonate
PubChem CID 146490
IUPAC Name [(2S)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES [O-][N+](=O)C1=CC=CC(=C1)S(=O)(=O)OC[C@@H]1CO1

Specifications

Melting Point 63°C to 66°C
Quantity 5 g
Beilstein 4704457
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methylene chloride and tetrahydrofuran. Insoluble in hexane.
Formula Weight 259.23
Percent Purity 98%
Chemical Name or Material Glycidyl (S)-(+)-3-nitrobenzenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.