missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl phenyl ether, 99%

Product Code. 10357210 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357210 250 g Glass bottle 250g
10606342 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10357210 Supplier Thermo Scientific Acros Supplier No. 135352500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-60-1 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00005133,MFCD31699959
InChI Key FQYUMYWMJTYZTK-UHFFFAOYNA-N
Synonym glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
PubChem CID 31217
ChEBI CHEBI:82367
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(OC1=CC=CC=C1)C1CO1

Specifications

Melting Point 3.5°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 245.0°C
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5295 to 1.5315
Quantity 250 g
Specific Gravity 1.1
Solubility Information Solubility in water: 2.4g/L(20°C). Other solubilities: miscible with ethanol
Viscosity 7 mPa.s (20°C)
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Glycidyl phenyl ether, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glycidyl phenyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.