missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Oleate-d5, TRC

Product Code. 30395381
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395381 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395381 Supplier Toronto Research Chemicals Supplier No. TRCG61593225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycidyl Oleate-d5
CAS 1426395-63-2
Purity Grade Notes GC
Molecular Formula C21 D5 H33 O3
Quantity 25 mg
Synonym (9Z)-9-Octadecenoic Acid 2-Oxiranylmethyl Ester-d5, Glycidyl Oleate D5 (Glycidyl D5), 9-Octadecenoic acid (9Z)-, 2-oxiranyl-2,3,3-d3-methyl-d2 ester, Oleic acid-glycidyl ester D5 (glycidyl D5)
InChI Formula InChI=1S/C21H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h9-10,20H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D
SMILES [2H]C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C1([2H])OC1([2H])[2H]
IUPAC Name [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] (Z)-octadec-9-enoate
Molecular Weight (g/mol) 343.56
Formula Weight 343.3135
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.