missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl Oleate-D5, TRC
Specifications
Specifications
| Chemical Name or Material | Glycidyl Oleate-d5 |
| CAS | 1426395-63-2 |
| Purity Grade Notes | GC |
| Molecular Formula | C21 D5 H33 O3 |
| Quantity | 2.5 mg |
| Synonym | (9Z)-9-Octadecenoic Acid 2-Oxiranylmethyl Ester-d5, Glycidyl Oleate D5 (Glycidyl D5), 9-Octadecenoic acid (9Z)-, 2-oxiranyl-2,3,3-d3-methyl-d2 ester, Oleic acid-glycidyl ester D5 (glycidyl D5) |
| InChI Formula | InChI=1S/C21H38O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h9-10,20H,2-8,11-19H2,1H3/b10-9-/i18D2,19D2,20D |
| SMILES | [2H]C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C1([2H])OC1([2H])[2H] |
| IUPAC Name | [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 343.56 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?