missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Myristate-d5 (1.0 mg/mL in isopropanol), TRC

Product Code. 30476269
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476269 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30476269 Supplier Toronto Research Chemicals Supplier No. TRCKIT83821X1ML
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1330180-72-7
Molecular Formula C17H27D5O3
Quantity 1 mL
Synonym Tetradecanoic Acid 2-Oxiranylmethyl-d5 Ester, Glycidyl-d5 Tetradecanoate, Myristic Acid Glycidyl-d5 Ester, NSC 404224-d5
InChI Formula InChI=1S/C17H32O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-17(18)20-15-16-14-19-16/h16H,2-15H2,1H3/i14D2,15D2,16D
SMILES O=C(OC([2H])([2H])C1([2H])OC1([2H])[2H])CCCCCCCCCCCCC
IUPAC Name [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] tetradecanoate
Molecular Weight (g/mol) 289.46
Formula Weight 289.2665
Concentration or Composition (by Analyte or Components) Glycidyl Myristate-d5 1 mg/mL
Chemical Name or Material Glycidyl Myristate-d5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.