missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Linoleate-d5, TRC

Product Code. 30240947
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240947 10 mg 10mg
30240873 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240947 Supplier Toronto Research Chemicals Supplier No. TRCG61572210MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246834-15-0
Molecular Formula C21 2H5 H31 O3
Molecular Weight (g/mol) 341.54
Synonym Glycidyl Linoleate D5 (Glycidyl D5), Linoleic acid-glycidyl ester D5 (glycidyl D5)
IUPAC Name [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] (9Z,12Z)-octadeca-9,12-dienoate
SMILES [2H]C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C1([2H])OC1([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h6-7,9-10,20H,2-5,8,11-19H2,1H3/b7-6-,10-9-/i18D2,19D2,20D
Formula Weight 341.2978
Chemical Name or Material Glycidyl Linoleate glycidyl-d5
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.