missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Linoleate-d5 (1.0 mg/mL in isopropanol), TRC

Product Code. 30473649
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473649 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473649 Supplier Toronto Research Chemicals Supplier No. TRCKIT83971X1ML
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1246834-15-0
Molecular Formula C21 2H5 H31 O3
Quantity 1 mL
Synonym Glycidyl Linoleate D5 (Glycidyl D5), Linoleic acid-glycidyl ester D5 (glycidyl D5)
InChI Formula InChI=1S/C21H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(22)24-19-20-18-23-20/h6-7,9-10,20H,2-5,8,11-19H2,1H3/b7-6-,10-9-/i18D2,19D2,20D
SMILES [2H]C([2H])(OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)C1([2H])OC1([2H])[2H]
IUPAC Name [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol) 341.54
Formula Weight 341.2978
Concentration or Composition (by Analyte or Components) Glycidyl Linoleate glycidyl-d5 1 mg/mL
Chemical Name or Material Glycidyl Linoleate glycidyl-d5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.