missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycidyl Laurate, TRC
Specifications
Specifications
| Chemical Name or Material | Glycidyl Laurate |
| CAS | 1984-77-6 |
| Molecular Formula | C15 H28 O3 |
| Quantity | 100 mg |
| Synonym | Dodecanoic Acid 2-Oxiranylmethyl Ester, Lauric Acid 2,3-Epoxypropyl Ester, 2,3-Epoxy-1-propanol Laurate, Glycidyl Dodecanoate, Lauric Acid Glycidyl Ester, NSC 406541 |
| InChI Formula | InChI=1S/C15H28O3/c1-2-3-4-5-6-7-8-9-10-11-15(16)18-13-14-12-17-14/h14H,2-13H2,1H3 |
| SMILES | CCCCCCCCCCCC(=O)OCC1CO1 |
| IUPAC Name | oxiran-2-ylmethyl dodecanoate |
| Molecular Weight (g/mol) | 256.38 |
| Formula Weight | 256.2038 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?