missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl Caprylate, TRC

Product Code. 30390258
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390258 100 mg
30401004 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390258 Supplier Toronto Research Chemicals Supplier No. TRCG615520100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24412-91-7
Molecular Formula C11H20O3
Molecular Weight (g/mol) 200.27
Synonym Octanoic Acid 2,3-Epoxypropyl Ester, Octanoic Acid Oxiranylmethyl Ester, 2,3-Epoxy-1-propanol Octanoate, 2,3-Epoxypropyl Octanoate
IUPAC Name oxiran-2-ylmethyl octanoate
SMILES CCCCCCCC(=O)OCC1CO1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H20O3/c1-2-3-4-5-6-7-11(12)14-9-10-8-13-10/h10H,2-9H2,1H3
Formula Weight 200.14
Chemical Name or Material Glycidyl Caprylate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.