missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 2-methylphenyl ether, tech. 85%

Product Code. 11308255
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11308255

Brand: Thermo Scientific Alfa Aesar L10136.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2210-79-9 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-79-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00037847
InChI Key KFUSXMDYOPXKKT-UHFFFAOYSA-N
Synonym 2-2-methylphenoxy methyl oxirane, o-cresyl glycidyl ether, glycidyl 2-methylphenyl ether, o-cresol glycidyl ether, glycidyl o-tolyl ether, araldite dy 023, o-kresol-glycidaether, 2-o-tolyloxy methyl oxirane, 1,2-epoxy-3-2-methylphenoxy propane, 1-2-methylphenoxy-2,3-epoxypropane
PubChem CID 16640
IUPAC Name 2-[(2-methylphenoxy)methyl]oxirane
SMILES CC1=CC=CC=C1OCC2CO2

Specifications

Density 1.079
Boiling Point 119°C to 120°C (6 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.529
Quantity 25 g
UN Number UN3082
Beilstein 4585
Formula Weight 164.21
Percent Purity 85%
Grade Technical
Chemical Name or Material Glycidyl 2-methylphenyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.