missing translation for 'onlineSavingsMsg'
Learn More

Glycidol (~90%), TRC

Product Code. 30402337
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402337 Supplier Toronto Research Chemicals Supplier No. TRCG6157031G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycidol
CAS 556-52-5
Molecular Formula C3 H6 O2
Quantity 1 g
Synonym Glycidol, Oxiran-2-ylmethanol, [(2RS)-Oxiran-2-yl]methanol, 2-Oxiranemethanol, 1-Propanol, 2,3-epoxy- (7CI,8CI), Glycidol (6CI), Oxiranemethanol (9CI), (RS)-Glycidol, (±)-2,3-Epoxy-1-propanol, (±)-Glycidol, 1,2-Epoxy-3-hydroxypropane, 1-Hydroxy-2,3-epoxypropane, 2,3-Epoxy-1-propanol, 2-(Hydroxymethyl)oxirane, 3-Hydroxy-1,2-epoxypropane, 3-Hydroxypropylene oxide, Allyl alcohol oxide, Epihydrin alcohol, Epiol OH, Glycide, Glycidol-Virantage VW 10200RP triblock copolymer, Glycidyl alcohol, NSC 46096, Oxiran-2-ylmethanol, Oxiranylmethanol, Racemic glycidol, dl-Glycidol
InChI Formula InChI=1S/C3H6O2/c4-1-3-2-5-3/h3-4H,1-2H2
SMILES OCC1CO1
IUPAC Name oxiran-2-ylmethanol
Molecular Weight (g/mol) 74.08
Formula Weight 74.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.