missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Triheptadecanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Glyceryl Triheptadecanoate |
| CAS | 2438-40-6 |
| Molecular Formula | C54H104O6 |
| Quantity | 1 g |
| Synonym | Heptadecanoic Acid 1,2,3-Propanetriyl Ester, Tri-heptadecanoin, Intarvin, Glycerol Triheptadecanoate, Glyceryl Margarate, Heptadecanoic Acid Triglyceride, Palmitic Acid Triglyderide, Triglyceride HdHdHd, Triheptadecanoin, Trimargarin, Trimargarine, Trimargaroylglycerol |
| InChI Formula | InChI=1S/C54H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-52(55)58-49-51(60-54(57)48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)50-59-53(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h51H,4-50H2,1-3H3 |
| SMILES | O=C(CCCCCCCCCCCCCCCC)OC(COC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O |
| IUPAC Name | 2,3-di(heptadecanoyloxy)propyl heptadecanoate |
| Molecular Weight (g/mol) | 849.4 |
| Formula Weight | 848.7833 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?