missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Triheptadecanoate, TRC

Product Code. 30400204
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400204 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400204 Supplier Toronto Research Chemicals Supplier No. TRCG6015652.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glyceryl Triheptadecanoate
CAS 2438-40-6
Molecular Formula C54H104O6
Quantity 2.5 g
Synonym Heptadecanoic Acid 1,2,3-Propanetriyl Ester, Tri-heptadecanoin, Intarvin, Glycerol Triheptadecanoate, Glyceryl Margarate, Heptadecanoic Acid Triglyceride, Palmitic Acid Triglyderide, Triglyceride HdHdHd, Triheptadecanoin, Trimargarin, Trimargarine, Trimargaroylglycerol
InChI Formula InChI=1S/C54H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-52(55)58-49-51(60-54(57)48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)50-59-53(56)47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h51H,4-50H2,1-3H3
SMILES O=C(CCCCCCCCCCCCCCCC)OC(COC(CCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCC)=O
IUPAC Name 2,3-di(heptadecanoyloxy)propyl heptadecanoate
Molecular Weight (g/mol) 849.4
Formula Weight 848.7833
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.