missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monooleate, MP Biomedicals

Product Code. 11412835 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412835 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412835 Supplier MP Biomedicals Supplier No. 0521101380
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 25496-72-4
Molecular Formula C21H40O4
MDL Number MFCD00067258,MFCD00042735,MFCD00069635
Quantity 100 g
Synonym monoelaidin, 1-monoelaidin, dl-alpha-monoolein, 2,3-dihydroxypropyl elaidate, glycerol monooleate, 9-octadecenoic acid, 2,3-dihydroxypropyl ester, e, 2,3-dihydroxypropyl 9e-octadec-9-enoate, 1-elaidoylglycerol, arlacel tm 186, glycerol-1-monooleate
InChI Key RZRNAYUHWVFMIP-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO
IUPAC Name 2,3-dihydroxypropyl (9Z)-octadec-9-enoate
Molecular Weight (g/mol) 356.55
PubChem CID 5364833
Formula Weight 356.55
Chemical Name or Material Glyceryl Monooleate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.