missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Monooleate, MP Biomedicals™

Product Code. 17831421 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17831421 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17831421 Supplier MP Biomedicals Supplier No. 0210488880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Releases free glycerol and oleic acid upon hydrolysis. Glyceryl Monooleate, MP Biomedicals is a surfactant that has been used in liquid crystal studies. Research shows that in the presence of Monoolein (1-Oleoyl-rac-glycerol), the penetration of the drug

Monoolein (1-Oleoyl-rac-glycerol) is used in the development of monoolein-based nanoparticulate liquid dispersions as possible vehicles for drug delivery.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-03-5
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.547
InChI Key RZRNAYUHWVFMIP-KTKRTIGZSA-N
Synonym monoolein, glyceryl monooleate, 2,3-dihydroxypropyl oleate, 1-monoolein, 1-oleoylglycerol, 1-oleoyl-rac-glycerol, glyceryl oleate, 1-glyceryl oleate, 1-monooleoylglycerol, glycerol 1-monooleate
PubChem CID 5283468
ChEBI CHEBI:75342
IUPAC Name 2,3-dihydroxypropyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(CO)O

Specifications

Melting Point 35°C (literature)
Quantity 100 mg
Solubility Information Soluble in chloroform (50mg/mL). Also soluble in hexane.
Formula Weight 356.55
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.