missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Iodide, TRC

Product Code. 30392084
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392084 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392084 Supplier Toronto Research Chemicals Supplier No. TRCG601495500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-iodopropane-1,2-diol
CAS 554-10-9
Molecular Formula C3 H7 I O2
Quantity 500 mg
InChI Formula InChI=1S/C3H7IO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
SMILES OCC(O)CI
IUPAC Name 3-iodopropane-1,2-diol
Molecular Weight (g/mol) 201.991
Formula Weight 201.949
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.