missing translation for 'onlineSavingsMsg'
Learn More

Glyceryl Behenate, TRC

Product Code. 30395854
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395854 100 mg
30397432 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395854 Supplier Toronto Research Chemicals Supplier No. TRCG601515100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18641-57-1
Molecular Formula C69 H134 O6
Molecular Weight (g/mol) 1059.8
Synonym Docosanoin, tri- (7CI,8CI), Behenic acid triglyceride, Compritol 888ATO, Compritol ATO 888, Glycerin tribehenate, Glycerin tribehenic acid ester, Glycerol tribehenate, Glyceryl behenate, Glyceryl tribehenate, Glyceryl tridocosanoate, Lipovol GTB, Pelemol GTB, Radia 7515, Syncrowax HRC, Syncrowax HRC-PA-MH, Syncrowax HRC-PA-RB, Syncrowax HRS, Tribehenin, Tribehenoylglycerol, Tridocosanoin
IUPAC Name 2,3-di(docosanoyloxy)propyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C69H134O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-52-55-58-61-67(70)73-64-66(75-69(72)63-60-57-54-51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)65-74-68(71)62-59-56-53-50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h66H,4-65H2,1-3H3
Formula Weight 1059.018
Chemical Name or Material Glyceryl Behenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.