missing translation for 'onlineSavingsMsg'
Learn More

Glycerophosphoric Acid Sodium Salt Hydate, Racemic, TRC

Product Code. 30138601
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138601 10 mg 10mg
30135589 50 mg 50mg
17987824 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138601 Supplier Toronto Research Chemicals Supplier No. TRCG60161010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C3H9O6P • x(Na) • y(H2O)
Molecular Weight (g/mol) 172.0722991802
Synonym rac-Glycerol 1-phosphate sodium salt hydrate, α-Glycerol phosphate sodium salt hydrate, DL-Glycerol-1-phosphate sodium salt hydrate, DL-Glycerol-3-phosphate sodium salt hydrate, Sodium 3-phosphoglycerate hydrate, Sodium DL-glycerophosphate hydrate
SMILES O=P(O)(O)OCC(O)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C3H9O6P/c4-1-3(5)2-9-10(6,7)8/h3-5H,1-2H2,(H2,6,7,8)
Chemical Name or Material Glycerophosphoric acid sodium salt hydate, Racemic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.