missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ultrapure, Spectrophotometric Grade

Product Code. 11348387
Click to view available options
Quantity:
100 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
100mL
4 L
This item is not returnable. View return policy

Product Code. 11348387

Brand: Thermo Scientific Alfa Aesar 032450.M1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production. It is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.26
Boiling Point 182°C (20 mm)
Flash Point 160°C (320°F)
Ignition Point 400°C (752°F)
Odor Odorless
Refractive Index 1.474
Quantity 4 x 1 L
Beilstein 635685
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 92.09
Vapor Pressure <0.1 hPa (0mm Hg) at 20°C
Grade Spectrophotometric
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.