missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Trilinoleate, TRC
Specifications
Specifications
| Chemical Name or Material | Glycerol Trilinoleate |
| CAS | 537-40-6 |
| Molecular Formula | C57 H98 O6 |
| Quantity | 100 mg |
| Synonym | 9,12-Octadecadienoic acid (9Z,12Z)-, 1,1',1''-(1,2,3-propanetriyl) ester, 9,12-Octadecadienoic acid (9Z,12Z)-, 1,2,3-propanetriyl ester (9CI), 9,12-Octadecadienoic acid (Z,Z)-, 1,2,3-propanetriyl ester, Linolein, tri- (6CI,7CI,8CI), Efaderma F, Glycerin trilinoleate, Glycerol trilinoleate, Glyceryl trilinoleate, LLL, LLL triacylglycerol, Linoleic acid triglyceride, Linoleic triglyceride, Triglyceride LLL, Trilinolein, Trilinoleoylglycerol, Trilinoleyl glyceride |
| InChI Formula | InChI=1S/C57H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16-21,25-30,54H,4-15,22-24,31-53H2,1-3H3/b19-16-,20-17-,21-18-,28-25-,29-26-,30-27- |
| SMILES | CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC |
| IUPAC Name | 2,3-bis[[(9Z,12Z)-octadeca-9,12-dienoyl]oxy]propyl (9Z,12Z)-octadeca-9,12-dienoate |
| Molecular Weight (g/mol) | 879.38 |
| Formula Weight | 878.7363 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?