missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Trilinoleate, TRC

Product Code. 30389701
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389701 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389701 Supplier Toronto Research Chemicals Supplier No. TRCG6015351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycerol Trilinoleate
CAS 537-40-6
Molecular Formula C57 H98 O6
Quantity 1 g
Synonym 9,12-Octadecadienoic acid (9Z,12Z)-, 1,1',1''-(1,2,3-propanetriyl) ester, 9,12-Octadecadienoic acid (9Z,12Z)-, 1,2,3-propanetriyl ester (9CI), 9,12-Octadecadienoic acid (Z,Z)-, 1,2,3-propanetriyl ester, Linolein, tri- (6CI,7CI,8CI), Efaderma F, Glycerin trilinoleate, Glycerol trilinoleate, Glyceryl trilinoleate, LLL, LLL triacylglycerol, Linoleic acid triglyceride, Linoleic triglyceride, Triglyceride LLL, Trilinolein, Trilinoleoylglycerol, Trilinoleyl glyceride
InChI Formula InChI=1S/C57H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h16-21,25-30,54H,4-15,22-24,31-53H2,1-3H3/b19-16-,20-17-,21-18-,28-25-,29-26-,30-27-
SMILES CCCCC\C=C/C\C=C/CCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C/C\C=C/CCCCC)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
IUPAC Name 2,3-bis[[(9Z,12Z)-octadeca-9,12-dienoyl]oxy]propyl (9Z,12Z)-octadeca-9,12-dienoate
Molecular Weight (g/mol) 879.38
Formula Weight 878.7363
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.