missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Solution, puriss p.a., 86 to 89% (T), Honeywell Riedel-de Haën™

Product Code. 15615700
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Poly bottle
Unit Size:
1 L
5 L
500mL
This item is not returnable. View return policy

Product Code. 15615700

Brand: Honeywell Riedel-de Haën 497825L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., 86-89% (T)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 20°C
Density 1.252g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Packaging Poly bottle
Linear Formula HOCH2CH(OH)CH2OH · aq
Refractive Index 1.454
Quantity 5 L
UN Number NONH for all modes of transport
Beilstein 635685
Formula Weight 92.09g/mol (anhydrous basis)
Percent Purity 86 to 89% (T)
Chemical Name or Material Glycerol solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.