missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Solution, Honeywell
£60.29 - £869.54
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.09 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | OCC(O)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15641480
|
Honeywell Riedel-de Haën
15524-1L |
1 L | |||||||
15671480
|
Honeywell Riedel-de Haën
15524-5L |
5 L | |||||||
15651480
|
Honeywell Riedel-de Haën
15524-25L |
25 L | |||||||
15691480
|
Honeywell Riedel-de Haën
15524-70KG |
70 kg | |||||||
Description
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Specifications
56-81-5 | |
160°C (320°F) | |
C3H8O3 | |
MFCD00004722 | |
635685 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
92.09g/mol (anhydrous basis) | |
puriss. |
1.252g/mL at 25°C | |
≤0.01% (as SO4) | |
HOCH2CH(OH)CH2OH · aq | |
NONH for all modes of transport | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
OCC(O)CO | |
92.09 | |
CHEBI:17754 | |
84 to 88% | |
Glycerol Solution, Meets analytical specification of Ph. Eur. BP |
Safety and Handling
missing translation for 'einecsNumber' : 200-289-5
missing translation for 'rtecsNumber' : MA8050000