missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, PlusOne™, Cytiva

Product Code. 11555005
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 56-81-5
Density 1.2g/mL (at 20°C)
Assay Percent Range 85%
Molecular Formula C3H8O3
MDL Number MFCD00004722
Quantity 1 L
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Formula Weight 92.09
Chemical Name or Material Glycerol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'