missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Monoleate, TRC

Product Code. 30003269
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003269 100 mg 100mg
30007887 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003269 Supplier Toronto Research Chemicals Supplier No. TRCG601540100MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25496-72-4
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.54
Synonym (Z)-9-Octadecenoic Acid Monoester with 1,2,3-Propanetriol, Olein, mono- (6CI,8CI), A 300, A 300 (ester), Adchem GMO, Agnique GMO, Ajax GMO, Aldo 40, Aldo MO, Aldo MO-FG, Alkamuls GMO 45LG, Arlacel 129, Atmer 1007, Atsurf 595, Canamex Glicepol 182, Capmul GMO 50, Crossential GMO, Dimodan GMO 90, Dimodan LSQK, Dremulse, Dur-Em 114, Dur-Em 204, EAR 300, Edenor GMO, Emalsy MO, Emalsy OL, Emalsy OL 100, Emalsy OL 100H, Emasol MO, Emasol MO 50, Emcol O, Emerest 2400, Emerest 2421, Emrite 6009, Emuldan RYLO-MG 90, Esterol 272, Excel O 95F, Excel O 95N, Excel O 95R, GMO 8903, GMOrphic 80, Glycerin monooleate, Glycerine monooleate, Glycerol monooleate, Glycerol oleate, Glyceromonooleate, Glyceryl monooleate, Glyceryl oleate, Glycolube 100, Harowax L 9, Kemester 2000, Kessco GMO, Loxiol G 10, Loxiol G 10V, Lumulse GMO, Mazol GMO, Monegyl 0100, Monoglyceryl oleate, Monomuls 90-018, Monomuls 90O18, Monoolein, Monooleoylglycerol, Nikkol MGO, OL 100, Oleic acid glycerol monoester, Oleic acid monoglyceride, Oleic monoglyceride, Oleylmonoglyceride, Olicine, Poem OL 200V, Polybatch AF 1085, Priolube 1407, Radiasurf 7149, Radiasurf 7150, Rheodol MO 50, Rheodol MO 60, Rikemal OL 100, Rikemal OL 100E, Rikemal XO 100, Rylo MG 19 Pharma, S 1096, S 1096R, S 1097, Sinnoester OGC, Sunsoft 8070, Sunsoft 8070V, Sunsoft O 30B, Sunsoft O 30S, Supeol, Synative ES-GMO, Tegin OV, XU 61518.10
SMILES O=C(OCC(O)CO)CCCCCCC/C=C\CCCCCCCC.O=C(OC(CO)CO)CCCCCCC/C=C\CCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h2*9-10,20,22-23H,2-8,11-19H2,1H3/b2*10-9-
Formula Weight 356.29
Chemical Name or Material Glyceryl Monooleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.