missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glycerol kinase Recombinant Rabbit Monoclonal Antibody (8S4T8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537862
This item is not returnable.
View return policy
Description
Positive Samples: 293T, U-87MG, Mouse testis, Rat liver, Rat testis, Rat kidney Immunogen sequence: IPVVKPSMPE TTALGAAMAA GAAEGVGVWS LEPEDLSAVT MERFEPQINA EESEIRYSTW KKAVMKSMGW VTTQSPESGD PSIFCSLPLG FFIVSSMVML IGARYISGIP.
The product of this gene belongs to the FGGY kinase family of proteins and encodes glycerol kinase. Glycerol kinase is a key enzyme in the regulation of glycerol uptake and metabolism. It catalyzes the phosphorylation of glycerol by ATP, yielding ADP and glycerol-3-phosphate. Defects in this gene are the cause of glycerol kinase deficiency (GKD). Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Glycerol kinase | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P32189, Q63060, Q64516 | |
| GK | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 8S4T8 | |
| Unconjugated | |
| GK | |
| ASTP; ATP:glycerol 3-phosphotransferase; ATP-stimulated glucocorticoid-receptor translocation promoter; ATP-stimulated glucocorticoid-receptor translocaton promoter; D930012N15Rik; EC 2.7.1.30; GK; GK 3; GK1; GK3P; GKD; GKl; GKP3; GKTB; glycerokinase; Glycerokinase 3; glycerol kinase; Glycerolkinase; Gyk | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14933, 2710, 79223 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction