missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Kinase Rabbit anti-Human, Mouse, Rat, Clone: 8S4T8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Glycerol Kinase |
|---|---|
| Clone | 8S4T8 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305296
|
Novus Biologicals
NBP3-15687-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18370116
|
Novus Biologicals
NBP3-15687-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycerol Kinase Monoclonal antibody specifically detects Glycerol Kinase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Glycerol Kinase | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase | |
| A synthetic peptide corresponding to a sequence within amino acids 450-559 of human Glycerol Kinase (P32189). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8S4T8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2710 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title