missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Glycerol Kinase Polyclonal specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | Glycerol Kinase |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:4000 - 1:20000 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | P04806 |
| Gene Alias | ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase |
| Gene Symbols | GK |
| Host Species | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?