missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Kinase Antibody [CoraFluor™ 1], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-78018CL1
This item is not returnable.
View return policy
Description
Glycerol Kinase Polyclonal antibody specifically detects Glycerol Kinase in Bacteria samples. It is validated for ELISA,Western Blot
Specifications
| Glycerol Kinase | |
| Polyclonal | |
| PBS | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase | |
| Glycerol Kinase [Cellulomonas species] (Uniprot: D0VZG4) | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 2710 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction