missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Kinase Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18431088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431088 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431088 Supplier Novus Biologicals Supplier No. NBP178018AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Glycerol Kinase Polyclonal antibody specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycerol Kinase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase
Host Species Goat
Immunogen Glycerol Kinase [Cellulomonas species] (Uniprot: D0VZG4)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2710
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.