missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol-d8, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Glycerol-d8 |
| CAS | 7325-17-9 |
| Molecular Formula | C3 D8 O3 |
| Quantity | 1 g |
| Synonym | 1,2,3-Propane-1,1,2,3,3-d5-triol-1,2,3-d3 (ACI), 1,2,3-Propane-1,1,2,3,3-d5-triol-d3 (9 CI), Glycerol-d8 (7 CI, 8 CI), 1,1,2,3,3-Pentadeuterio-1,2,3-trideuteriooxypropane, Glycerol-D8 |
| InChI Formula | InChI=1 S/C3H8O3/c4-1-3(6)2-5/h3-6 H,1-2H2/i1D2,2D2,3 D,4 D,5 D,6 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])(O[2 H])C([2 H])([2 H])O[2 H] |
| IUPAC Name | 1,1,2,3,3-pentadeuterio-1,2,3-trideuteriooxypropane |
| Molecular Weight (g/mol) | 100.1431 |
| Formula Weight | 100.0976 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction