missing translation for 'onlineSavingsMsg'
Learn More

Glycerol-d5 1,3-Dipalmitate, TRC

Product Code. 30395319
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395319 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30395319 Supplier Toronto Research Chemicals Supplier No. TRCG5984125MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycerol-d5 1,3-Dipalmitate
CAS 65615-82-9
Molecular Formula C35D5H63O5
Quantity 5 mg
Synonym 1,3-Dipalmitin-d5, 1,3-Dipalmitoylglycerin-d5, 1,3-Dipalmitoylglycerol-d5, Glycerol-d5 1,3-Dihexadecanoate, Hexadecanoic-d5 Acid 2-Hydroxy-1,3-propanediyl Ester
InChI Formula InChI=1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3/i31D2,32D2,33D
SMILES [2H]C([2H])(OC(=O)CCCCCCCCCCCCCCC)C([2H])(O)C([2H])([2H])OC(=O)CCCCCCCCCCCCCCC
IUPAC Name (1,1,2,3,3-pentadeuterio-3-hexadecanoyloxy-2-hydroxy-propyl) hexadecanoate
Molecular Weight (g/mol) 573.94
Formula Weight 573.538
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.