missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Anhydrous, Ultrapure Bioreagent, J.T. Baker™

Product Code. p-5383267 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15548224

Brand: Avantor J.T.Baker 404300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For enzyme stabilization, freezing and electrophoresis buffers

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Flash Point 199.00°C
Packaging HDPE Pressure Bottom Poly Bottle
Quantity 500 mL
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Grade Ultrapure Bioreagent
Chemical Name or Material Glycerol, Anhydrous
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.