missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
Each
Specifications
Specifications
| Density | 1.257g/mL at 25°C (typical) |
| Flash Point | 199.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Quantity | 4 L |
| Specific Gravity | 1L = 1.26kg |
| Formula Weight | 92.1 |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | Glycerol, Anhydrous |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction