missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, for analysis, 86-88% wt% aqueous solution

Product Code. 10665521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10665521

Brand: Thermo Scientific Acros 295600025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5, 7732-18-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.0°C
Color Colorless
Density 1.2270g/mL
Boiling Point 290.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4520 to 1.4750
Quantity 2.5 L
Beilstein 01, 502
Merck Index 15, 4520
Specific Gravity 1.227
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform and in, fixed and volatile oils
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 86.0 to 88.0%
Grade Analysis
Physical Form Viscous Liquid
Chemical Name or Material Glycerol, 86 to 88% wt% Aqueous Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.