missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS, 99.5+%

Product Code. 11398078
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398078 4 L 4 L
11405190 500 mL 500mL
11308088 1 L 1 L
11318088 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11398078 Supplier Thermo Scientific Alfa Aesar Supplier No. 036646.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26
Boiling Point 182°C (20 mm)
Flash Point 160°C (320°F)
Ignition Point 400°C (752°F)
Assay Percent Range >99.5%
Refractive Index 1.474
Quantity 4 L
Beilstein 635685
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol,ethanol and 2-propanol. Immiscible in ether,benzene and chloroform.
Formula Weight 92.09
Vapor Pressure <0.1 hPa (0mm Hg) at 20°C
Grade ACS
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.