missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%, pure, synthetic

Product Code. 10716432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
100mL
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10716432 1 L Glass bottle 1 L
10001841 100 mL Glass bottle 100mL
10244991 2.5 L Glass bottle 2.5 L
11994631 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10716432 Supplier Thermo Scientific Acros Supplier No. 332030010
  • ONLINE EXCLUSIVE PRICE
    £270.00
    £249.00 / 1 L
    Save £21.00
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.0°C
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4716
Quantity 1 L
Beilstein 01,502
Merck Index 15,452
Specific Gravity 1.261
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 99+%
Grade Pure
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.