missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99% max., Molecular biology reagent grade, MP Biomedicals™

Product Code. 11476441 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL

Product Code. 11476441

Brand: MP Biomedicals 0219399680

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 56-81-5
Color Colorless
Density 1.2613g/cm3 at 20°C
Molecular Formula C3H8O3
MDL Number MFCD00004722
Quantity 100 mL
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES OCC(O)CO
IUPAC Name propane-1,2,3-triol
Molecular Weight (g/mol) 92.09
PubChem CID 753
ChEBI CHEBI:17754
Formula Weight 92.09
Percent Purity 99% max.
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.