missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15658490
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
4 L
4 x 4 L
Packaging:
Glass bottle
Poly bottle
Unit Size:
1 L
2 L
4 L
500mL
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15658490 4 L Poly bottle 4 L
15667890 500 mL Poly bottle 500mL
15637890 1 L Poly bottle 1 L
15647890 2 L Poly bottle 2 L
15657890 4 x 4 L Glass bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15658490 Supplier Honeywell-Riedel-de Haen Supplier No. G78934L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.25g/mL
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Ignition Point 698°F
Odor Odorless
Packaging Poly bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 4 L
UN Number NONH for all modes of transport
Beilstein 635685
Solubility Information Completely miscible in water
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.