missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2 L
4 L
4 x 4 L
Packaging:
Glass bottle
Poly bottle
Unit Size:
1 L
2 L
4 L
500mL
Pack of 4
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Melting Point | 18°C |
| Color | Colorless |
| Density | 1.25g/mL |
| pH | Neutral |
| Boiling Point | 290°C (1013 hPa) |
| Flash Point | 160°C (320°F) |
| Ignition Point | 698°F |
| Odor | Odorless |
| Packaging | Glass bottle |
| Linear Formula | HOCH2CH(OH)CH2OH |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction