missing translation for 'onlineSavingsMsg'
Learn More
Learn More
glycerol-3-phosphate permease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87524
This item is not returnable.
View return policy
Description
glycerol-3-phosphate permease Polyclonal specifically detects glycerol-3-phosphate permease in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| glycerol-3-phosphate permease | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ22340, G-3-P permease, G-3-P transporter, G3PPgene similar to glycerol-3-phosphate permease10Glycerol-3-phosphate permease, glycerol-3-phosphate transporter, solute carrier family 37 (glycerol-3-phosphate transporter), member 1, Solute carrier family 37 member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 54020 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC37A1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IVKGELHKYCTAWDEADVRFSSQNRKSGSAAPHQLPDNETDCGWAPFDKNNY | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction