missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol 3 Phosphate Dehydrogenase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97069-50ul
This item is not returnable.
View return policy
Description
Glycerol 3 Phosphate Dehydrogenase Polyclonal antibody specifically detects Glycerol 3 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Glycerol 3 Phosphate Dehydrogenase | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 μL/mg of lysate | |
| EC 1.1.1, EC 1.1.1.8, FLJ26652, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase 1 (soluble), glycerol-3-phosphate dehydrogenase, soluble, GPD-C, GPDH-C | |
| Produced in rabbits immunized with purified, recombinant Human Glycerol 3 Phosphate Dehydrogenase (Accession#: P21695; Met1-Met349) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2819 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction