missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol 3 Phosphate Dehydrogenase Antibody (104), DyLight 650, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Glycerol 3 Phosphate Dehydrogenase Monoclonal specifically detects Glycerol 3 Phosphate Dehydrogenase in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Glycerol 3 Phosphate Dehydrogenase |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 104 |
| Conjugate | DyLight 650 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.1.1, EC 1.1.1.8, FLJ26652, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase 1 (soluble), glycerol-3-phosphate dehydrogenase, soluble, GPD-C, GPDH-C |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Glycerol 3 Phosphate Dehydrogenase (P21695; Met1-Met349). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?