missing translation for 'onlineSavingsMsg'
Learn More

Glycerol 3 Phosphate Dehydrogenase Antibody (104), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18362541 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362541 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362541 Supplier Novus Biologicals Supplier No. NBP290308AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Glycerol 3 Phosphate Dehydrogenase Monoclonal specifically detects Glycerol 3 Phosphate Dehydrogenase in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycerol 3 Phosphate Dehydrogenase
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1, EC 1.1.1.8, FLJ26652, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase 1 (soluble), glycerol-3-phosphate dehydrogenase, soluble, GPD-C, GPDH-C
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Glycerol 3 Phosphate Dehydrogenase (P21695; Met1-Met349).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2819
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.