missing translation for 'onlineSavingsMsg'
Learn More

Glycerol 2-Methyl Ether, TRC

Product Code. 30401003
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401003 250 mg 250mg
30406412 100 mg -
30394095 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401003 Supplier Toronto Research Chemicals Supplier No. TRCG601550250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761-06-8
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
Synonym Glycerol 2-Monomethyl Ether, 2-O-Methylglycerol, 2-Methoxy-1,3-propanediol
IUPAC Name 2-methoxypropane-1,3-diol
SMILES OCC(OC)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H10O3/c1-7-4(2-5)3-6/h4-6H,2-3H2,1H3
Formula Weight 106.063
Chemical Name or Material Glycerol 2-Methyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.