missing translation for 'onlineSavingsMsg'
Learn More

Glycerol 1-Acetate (Technical grade), TRC

Product Code. 17968574
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17968574 250 mg 250mg
17990075 1 g 1g
17967324 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17968574 Supplier Toronto Research Chemicals Supplier No. TRCG601570250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-61-6
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
Synonym (±)-2,3-Dihydroxypropyl Acetate, 2,3-Dihydroxypropyl Acetate, Glycerine 1-Acetate, Glycerol 1-Monoacetate, Glycerol alpha-Monoacetate, Glyceryl 1-Monoacetate, NSC 35010, alpha-Monoacetin, 1-Monoacetin
IUPAC Name 2,3-dihydroxypropyl acetate
SMILES CC(=O)OCC(O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H10O4/c1-4(7)9-3-5(8)2-6/h5-6,8H,2-3H2,1H3
Formula Weight 134.0579
Chemical Name or Material Glycerol 1-Acetate (Technical grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.