missing translation for 'onlineSavingsMsg'
Learn More

Glycerol 1,3-Dipalmitate, TRC

Product Code. 30395545
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395545 100 mg
30403724 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395545 Supplier Toronto Research Chemicals Supplier No. TRCG598410100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-52-3
Molecular Formula C35H68O5
Molecular Weight (g/mol) 568.91
Synonym 1,3-Dipalmitin, 1,3-Dipalmitoylglycerin, 1,3-Dipalmitoylglycerol, Glycerol 1,3-Dihexadecanoate, Hexadecanoic Acid, 2-hydroxy-1,3-propanediyl Ester
IUPAC Name (3-hexadecanoyloxy-2-hydroxypropyl) hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3
Formula Weight 568.5067
Chemical Name or Material Glycerol 1,3-Dipalmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.