missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, MP Biomedicals™

Product Code. 11495255 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495255 4 L 4 L
11475255 500 mL 500mL
11485255 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495255 Supplier MP Biomedicals Supplier No. 0219956604
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in sample prep and gel formation for polyacrylamide gel electrophoresis. Glycerin, MP Biomedicals (5-10%) increases the density of a sample so that it will layer at the bottom of a sample well. It is also used in casting gradient gels and as a protei

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 20°C (literature)
Density 1.25g/mL (Lit.)
Boiling Point 182°C (20 mmHg (literature))
Ignition Residue ≤0.01%
Refractive Index n20/D 1.474 (Lit.)
Quantity 4 L
Specific Gravity ≥1.249
Vapor Density 3.1 (vs air) (Lit.)
Formula Weight 92.09g/mol
Vapor Pressure <1mmHg (20°C) (Lit.)
Percent Purity ≥99%
Grade USP
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.