missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, 100% Vegetable Based, U.S.P., J.T. Baker™

Product Code. 15507984
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Flash Point 199.00°C
Packaging Poly Pail
Quantity 19 L
Specific Gravity 1L = 1.26kg
Formula Weight 92.1
Percent Purity 100% Vegetable Based
Grade USP
Chemical Name or Material Glycerin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'