missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLYATL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569588
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
Catalyzes the conjugation of long-chain fatty acyl-CoA thioester and glycine to produce long-chain N-(fatty acyl)glycine, an intermediate in the primary fatty acid amide biosynthetic pathway. [UniProt].
Specifications
| GLYATL3 | |
| Polyclonal | |
| Unconjugated | |
| GLYATL3 | |
| Acyl-CoA:glycine-N-acyltransferase-like protein 3; bA28H17.2; C6orf140; EG435528; GLYATL3; Glycine N-acyltransferase-like protein 3; glycine N-acyltransferase-like protein 3-like; glycine-N-acyltransferase like 3; glycine-N-acyltransferase-like 3; Glycine-N-acyltransferase-like protein 3; Gm5683; LOW QUALITY PROTEIN: glycine N-acyltransferase-like protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 389396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5SZD4 | |
| GLYATL3 | |
| synthetic peptide directed towards the N terminal region of human GLYATL3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction